missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Luc7-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Luc7-like protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7504-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_006098.2 | |
Luc7-like protein 1 | |
Research Use Only | |
LUC7L3 | |
Recombinant Protein | |
E. coli | |
MISAAQLLDE LMGRDRNLAP DEKRSNVRWD HESVCKYYLC GFCPAELFTN TRSDLGPCEK IHDENLRKQY EKSSRFMKV | |
Greater than 90% as determined by SDS-PAGE. |
51747 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |