missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lutropin-choriogonadotropic hormone receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10085-ma-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000224.2 | |
Lutropin-choriogonadotropic hormone receptor Protein | |
Research Use Only | |
Lhcgr | |
Recombinant Protein | |
Mammal | |
EALCPEPCNC VPDGALRCPG PTAGLTRLSL AYLPVKVIPS QAFRGLNEVI KIEISQIDSL ERIEANAFDN LLNLSEILIQ NTKNLRYIEP GAFINLPRLK YLSICNTGIR KFPDVTKVFS SESNFILEIC DNLHITTIPG NAFQGMNNES VTLKLYGNGF EEVQSHAFNG TTLTSLELKE NVHLEKMHNG AFRGATGPKT LDISSTKLQA LPSYGLESIQ RLIATSSYSL KKLPSRETFV NLLEATLTYP SHCCAFRNLP TKEQNFSHSI SENFSKQCES TVRKVNNKTL YSSMLAESEL SGWDYEYGFC LPKTPRCAPE PDAFNPCEDI MGYDFL | |
Greater than 90% as determined by SDS-PAGE. |
3973 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |