missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Ly6/PLAUR domain-containing protein 6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7780-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001182614.1 | |
Ly6 / PLAUR domain-containing protein 1 | |
Research Use Only | |
LYPD6 | |
Recombinant Protein | |
E. coli | |
AQSRDFTVKD IIYLHPSTTP YPGGFKCFTC EKAADNYECN RWAPDIYCPR ETRYCYTQHT MEVTGNSISV TKRCVPLEEC LSTGCRDSEH EGHKVCTSCC EGNICNLPLP RNETDATFAT TSPINQTNGH PRCMSVIVSC LWLWLGLML | |
Greater than 90% as determined by SDS-PAGE. |
130574 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |