missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAG / GMA / Siglec-4 Protein
A DNA sequence encoding the Homo sapiens (Human) MAG / GMA / Siglec-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6332-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001186145.1 | |
MAG / GMA / Siglec-4 Protein | |
Research Use Only | |
MAG | |
Recombinant Protein | |
E. coli | |
GHWGAWMPSS ISAFEGTCVS IPCRFDFPDE LRPAVVHGVW YFNSPYPKNY PPVVFKSRTQ VVHESFQGRS RLLGDLGLRN CTLLLSNVSP ELGGKYYFRG DLGGYNQYTF SEHSVLDIVN TPNIVVPPEV VAGTEVEVSC MVPDNCPELR PELSWLGHEG LGEPAVLGRL REDEGTWVQV SLLHFVPTRE ANGHRLGCQA SFPNTTLQFE GYASMDVKYP PVIVEMNSSV EAIEGSHVSL LCGADSNPPP LLTWMRDGTV LREAVAESLL LELEEVTPAE DGVYACLAEN AYGQDNRTVG LSVMYAPWKP TVNGTMVAVE GETVSILCST QSNPDPILTI FKEKQILSTV IYESELQLEL PAVSPEDDGE YWCVAENQYG QRATAFNLSV EFAPVLLLES HCAAARDTVQ CLCVVKSNPE PSVAFELPSR NVTVNESERE FVYSERSGLV LTSILTLRGQ AQAPPRVICT ARNLYGAKSL ELPFQGAHRL MWAKIGP | |
Greater than 90% as determined by SDS-PAGE. |
4099 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |