missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MART-1 / MLANA Protein
A DNA sequence encoding the Homo sapiens (Human) MART-1 / MLANA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7586-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005502.1 | |
MART-1 / MLANA Protein | |
Research Use Only | |
MLANA | |
Recombinant Protein | |
Yeast | |
MPREDAHFIY GYPKKGHGHS YTTAEEAAGI GILTVILGVL LLIGCWYCRR RNGYRALMDK SLHVGTQCAL TRRCPQEGFD HRDSKVSLQE KNCEPVVPNA PPAYEKLSAE QSPPPYSP | |
Greater than 90% as determined by SDS-PAGE. |
2315 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |