missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-1F Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1F, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9291-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001288201.1 | |
Metallothionein-1F Protein | |
Research Use Only | |
MT1F | |
Recombinant Protein | |
Yeast | |
MDPNCSCAAG VSCTCAGSCK CKECKCTSCK KSCCSCCPVG CSKCAQGCVC KGASEKCSC | |
Greater than 90% as determined by SDS-PAGE. |
4494 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |