missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methionine synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Methionine synthase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8028-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000245.2 | |
Methionine synthase Protein | |
Research Use Only | |
MTR | |
Recombinant Protein | |
E. coli | |
SLKERRYLPL SQARKSGFQM DWLSEPHPVK PTFIGTQVFE DYDLQKLVDY IDWKPFFDVW QLRGKYPNRG FPKIFNDKTV GGEARKVYDD AHNMLNTLIS QKKLRARGVV GFWPAQSIQD DIHLYAEAAV PQAAEPIATF YGLRQQAEKD SASTEPYYCL SDFIAPLHSG IRDYLGLFAV ACFGVEELSK AYEDDGDDYS SIMVKALGDR LAEAFAEELH ERVRRELWAY CGSEQLDVAD LRRLRYKGIR PAPGYPSQPD HTEKLTMWRL ADIEQSTGIR LTESLAMAPA SAVSGLYFSN LKSKYFAVGK ISKDQVEDYA LRKNISVAEV EKWLGPILGY DTD | |
Greater than 90% as determined by SDS-PAGE. |
4548 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |