missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MICOS complex subunit MIC19 Protein
A DNA sequence encoding the Homo sapiens (Human) MICOS complex subunit MIC19, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8122-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001304106.1 | |
MICOS complex subunit MIC19 Protein | |
Research Use Only | |
CHCHD3 | |
Recombinant Protein | |
E. coli | |
MGGTTSTRRV TFEADENENI TVVKGIRLSE NVIDRMKESS PSGSKSQRYS GAYGASVSDE ELKRRVAEEL ALEQAKKESE DQKRLKQAKE LDRERAAANE QLTRAILRER ICSEEERAKA KHLARQLEEK DRVLKKQDAF YKEQLARLEE RSSEFYRVTT EQYQKAAEEV EAKFKRYESH PVCADLQAKI LQCYRENTHQ TLKCSALATQ YMHCVNHAKQ SMLEKGG | |
Greater than 80% as determined by SDS-PAGE. |
54927 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |