missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKRN3 Protein
A DNA sequence encoding the Homo sapiens (Human) MKRN3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7557-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005655.1 | |
MKRN3 Protein | |
Research Use Only | |
MKRN3 | |
Recombinant Protein | |
E. coli | |
MEEPAAPSEA HEAAGAQAGA EAAREGVSGP DLPVCEPSGE SAAPDSALPH AARGWAPFPV APVPAHLRRG GLRPAPASGG GAWPSPLPSR SSGIWTKQII CRYYIHGQCK EGENCRYSHD LSGRKMATEG GVSPPGASAG GGPSTAAHIE PPTQEVAEAP PAASSLSLPV IGSAAERGFF EAERDNADRG AAGGAGVESW ADAIEFVPGQ PYRGRWVASA PEAPLQSSET ERKQMAVGSG LRFCYYASRG VCFRGESCMY LHGDICDMCG LQTLHPMDAA QREEHMRACI EAHEKDMELS FAVQRGMDKV CGICMEVVYE KANPNDRRFG ILSNCNHSFC IRCIRRWRSA RQFENRIVKS CPQCRVTSEL VIPSEFWVEE EEEKQKLIQQ YKEAMSNKAC RYFAEGRGNC PFGDTCFYKH EYPEGWGDEP PGPGGGSFSA YWHQLVEPVR MGEGNMLYKS IKKELVVLRL ASLLFKRFLS LRDELPFSED QWDLLHYELE EYFNLIL | |
Greater than 90% as determined by SDS-PAGE. |
7681 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |