missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Monoglyceride lipase Protein
A DNA sequence encoding the Homo sapiens (Human) Monoglyceride lipase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6358-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001003794.1 | |
Monoglyceride lipase Protein | |
Research Use Only | |
Mgll | |
Recombinant Protein | |
E. coli | |
MPEESSPRRT PQSIPYQDLP HLVNADGQYL FCRYWKPTGT PKALIFVSHG AGEHSGRYEE LARMLMGLDL LVFAHDHVGH GQSEGERMVV SDFHVFVRDV LQHVDSMQKD YPGLPVFLLG HSMGGAIAIL TAAERPGHFA GMVLISPLVL ANPESATTFK VLAAKVLNLV LPNLSLGPID SSVLSRNKTE VDIYNSDPLI CRAGLKVCFG IQLLNAVSRV ERALPKLTVP FLLLQGSADR LCDSKGAYLL MELAKSQDKT LKIYEGAYHV LHKELPEVTN SVFHEINMWV SQRTATAGTA SPP | |
Greater than 80% as determined by SDS-PAGE. |
11343 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |