missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MSRB3 Protein
A DNA sequence encoding the Homo sapiens (Human) MSRB3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7810-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001026849.1 | |
MSRB3 Protein | |
Research Use Only | |
MSRB3 | |
Recombinant Protein | |
E. coli | |
MSAFNLLHLV TKSQPVALRA CGLPSGSCRD KKNCKVVFSQ QELRKRLTPL QYHVTQEKGT ESAFEGEYTH HKDPGIYKCV VCGTPLFKSE TKFDSGSGWP SFHDVINSEA ITFTDDFSYG MHRVETSCSQ CGAHLGHIFD DGPRPTGKRY CINSAALSFT PADSSGTAEG GSGVASPAQA DKAEL | |
Greater than 80% as determined by SDS-PAGE. |
253827 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |