missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-acetylgalactosamine-6-sulfatase Protein
A DNA sequence encoding the Homo sapiens (Human) N-acetylgalactosamine-6-sulfatase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6066-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000503.1 | |
N-acetylgalactosamine-6-sulfatase Protein | |
Research Use Only | |
GALNS | |
Recombinant Protein | |
E. coli | |
APQPPNILLL LMDDMGWGDL GVYGEPSRET PNLDRMAAEG LLFPNFYSAN PLCSPSRAAL LTGRLPIRNG FYTTNAHARN AYTPQEIVGG IPDSEQLLPE LLKKAGYVSK IVGKWHLGHR PQFHPLKHGF DEWFGSPNCH FGPYDNKARP NIPVYRDWEM VGRYYEEFPI NLKTGEANLT QIYLQEALDF IKRQARHHPF FLYWAVDATH APVYASKPFL GTSQRGRYGD AVREIDDSIG KILELLQDLH VADNTFVFFT SDNGAALISA PEQGGSNGPF LCGKQTTFEG GMREPALAWW PGHVTAGQVS HQLGSIMDLF TTSLALAGLT PPSDRAIDGL NLLPTLLQGR LMDRPIFYYR GDTLMAATLG QHKAHFWTWT NSWENFRQGI DFCPGQNVSG VTTHNLEDHT KLPLIFHLGR DPGERFPLSF ASAEYQEALS RITSVVQQHQ EALVPAQPQL NVCNWAVMNW APPGCEKLGK CLTPPESIPK KCLWSH | |
Greater than 90% as determined by SDS-PAGE. |
2588 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |