missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAD-dependent protein deacetylase sirtuin-6 Protein
A DNA sequence encoding the Homo sapiens (Human) NAD-dependent protein deacetylase sirtuin-6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7987-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001180214.1 | |
NAD-dependent protein deacetylase sirtuin-6 Protein | |
Research Use Only | |
SIRT6 | |
Recombinant Protein | |
E. coli | |
SVNYAAGLSP YADKGKCGLP EIFDPPEELE RKVWELARLV WQSSSVVFHT GAGISTASGI PDFRGPHGVW TMEERGLAPK FDTTFESARP TQTHMALVQL ERVGLLRFLV SQNVDGLHVR SGFPRDKLAE LHGNMFVEEC AKCKTQYVRD TVVGTMGLKA TGRLCTVAKA RGLRACRGEL RDTILDWEDS LPDRDLALAD EASRNADLSI TLGTSLQIRP SGNLPLATKR RGGRLVIVNL QPTKHDRHAD LRIHGYVDEV MTRLMKHLGL EIPAWDGPRV LERALPPLPR PPTPKLEPKE ESPTRINGSI PAGPKQEPCA QHNGSEPASP KRERPTSPAP HRPPKRVKAK AVPS | |
Greater than 80% as determined by SDS-PAGE. |
51548 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |