missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEDD8 Protein
A DNA sequence encoding the Homo sapiens (Human) NEDD8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7554-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_006147.1 | |
NEDD8 Protein | |
Research Use Only | |
NEDD8 | |
Recombinant Protein | |
E. coli | |
MLIKVKTLTG KEIEIDIEPT DKVERIKERV EEKEGIPPQQ QRLIYSGKQM NDEKTAADYK ILGGSVLHLV LALRGG | |
Greater than 80% as determined by SDS-PAGE. |
4738 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |