missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NR2F6 Protein
A DNA sequence encoding the Homo sapiens (Human) NR2F6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6434-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005225.2 | |
NR2F6 Protein | |
Research Use Only | |
NR2F6 | |
Recombinant Protein | |
E. coli | |
MAMVTGGWGG PGGDTNGVDK AGGYPRAAED DSASPPGAAS DAEPGDEERP GLQVDCVVCG DKSSGKHYGV FTCEGCKSFF KRSIRRNLSY TCRSNRDCQI DQHHRNQCQY CRLKKCFRVG MRKEAVQRGR IPHSLPGAVA ASSGSPPGSA LAAVASGGDL FPGQPVSELI AQLLRAEPYP AAAGRFGAGG GAAGAVLGID NVCELAARLL FSTVEWARHA PFFPELPVAD QVALLRLSWS ELFVLNAAQA ALPLHTAPLL AAAGLHAAPM AAERAVAFMD QVRAFQEQVD KLGRLQVDSA EYGCLKAIAL FTPDACGLSD PAHVESLQEK AQVALTEYVR AQYPSQPQRF GRLLLRLPAL RAVPASLISQ LFFMRLVGKT PIETLIRDML LSGSTFNWPY GSGQ | |
Greater than 90% as determined by SDS-PAGE. |
2063 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |