missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NREP Protein

A cDNA sequence encoding the NREP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12879-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15951829

  • 1600.00€ / 100µg

Spedizione stimata: 31-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

9315
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
NREP
Human
His
The NREP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
NREP Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVYYPEL FVWVSQEPFP NKDMEGRLPK GRLPVPKEVN RKKNDETNAA SLTPLGSSEL RSPRISYLHF F
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali