missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Odorant-binding protein 2a Protein
A DNA sequence encoding the Homo sapiens (Human) Odorant-binding protein 2a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8123-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001280118.1 | |
Odorant-binding protein 2a Protein | |
Research Use Only | |
OBP2A | |
Recombinant Protein | |
E. coli | |
LSFTLEEEDI TGTWYVKAMV VDKDFPEDRR PRKVSPVKVT ALGGGNLEAT FTFMREDRCI QKKILMRKTE EPGKFSAYGG RKLIYLQELP GTDDYVFYCK DQRRGGLRYM GKLVGRNPNT NLEALEEFKK LVQHKGLSEE DIFMPLQTGS CVLEH | |
Greater than 90% as determined by SDS-PAGE. |
29991 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |