missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oncostatin M / OSM Protein
Recombinant Protein
Marca: enQuireBio™ QP5367-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001306037.1 | |
Oncostatin M / OSM Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
TF-1 cell proliferation was significantly induced by less than 4 ng/ml of Human Oncostatin M / OSM (minimum activity of at least 2.5 x 10^5 units/mg). | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
5008 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
OSM | |
Recombinant Protein | |
E. coli | |
MAAIGSCSKE . YRVLLGQLQ K. QTDLMQDT SR. LLDPYIR IQG. LDVPKL REHC. RERPG AFPSE. ETLR GLGRRG. FLQ TLNATLGCVL HRLADLE. QR LPKAQDLE. R SGLNIEDLE. KLQMARPNIL . GLRNNIYCM A. QLLDNSDT AE. PTKAGRG ASQ. PPTPTP ASDA. FQRKL EGCRFLHGYH RFMHS. VGRV FSKWGE. SPN RSRRHSP. HQ ALRKGVRR |