missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Osteoprotegerin / TNFRSF11B Protein
A DNA sequence encoding the Human Osteoprotegerin / TNFRSF11B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10426-ye-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002537.3 | |
Osteoprotegerin / TNFRSF11B Protein | |
Research Use Only | |
TNFRSF11B | |
The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL). | |
Human | |
FC | |
0.2um filtered 20 mM PBS, pH 6.0, 150 mM NaCl, 0.02 % Tween-80, lyophilized |
4982 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
Yeast | |
ETFPPKYLHY DEETSHQLLC DKCPPGTYLK QHCTAKWKTV CAPCPDHYYT DSWHTSDECL YCSPVCKELQ YVKQECNRTH NRVCECKEGR YLEIEFCLKH RSCPPGFGVV QAGTPERNTV CKRCPDGFFS NETSSKAPCR KHTNCSVFGL LLTQKGNATH DNICSGNSES TQKCGIDVTL EPKSSDKTHT CPPCPAPEFE GAPSVFLFPP KPKDTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPTPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNG QPENNYKTTP PVLDSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GK | |
Greater than 95% as determined by SDS-PAGE and HPLC. |