missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p38 delta / MAPK13 Protein
A DNA sequence encoding the Homo sapiens (Human) p38 delta / MAPK13, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6339-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002745.1 | |
p38 delta / MAPK13 Protein | |
Research Use Only | |
MAPK13 | |
Recombinant Protein | |
E. coli | |
MSLIRKKGFY KQDVNKTAWE LPKTYVSPTH VGSGAYGSVC SAIDKRSGEK VAIKKLSRPF QSEIFAKRAY RELLLLKHMQ HENVIGLLDV FTPASSLRNF YDFYLVMPFM QTDLQKIMGM EFSEEKIQYL VYQMLKGLKY IHSAGVVHRD LKPGNLAVNE DCELKILDFG LARHADAEMT GYVVTRWYRA PEVILSWMHY NQTVDIWSVG CIMAEMLTGK TLFKGKDYLD QLTQILKVTG VPGTEFVQKL NDKAAKSYIQ SLPQTPRKDF TQLFPRASPQ AADLLEKMLE LDVDKRLTAA QALTHPFFEP FRDPEEETEA QQPFDDSLEH EKLTVDEWKQ HIYKEIVNFS PIARKDSRRR SGMKL | |
Greater than 90% as determined by SDS-PAGE. |
5603 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |