missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human P4HB / ERBA2L Protein
A DNA sequence encoding the Homo sapiens (Human) P4HB / ERBA2L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8314-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000909.2 | |
P4HB / ERBA2L Protein | |
Research Use Only | |
P4hb | |
Recombinant Protein | |
E. coli | |
LRKSNFAEAL AAHKYLLVEF YAPWCGHCKA LAPEYAKAAG KLKAEGSEIR LAKVDATEES DLAQQYGVRG YPTIKFFRNG DTASPKEYTA GREADDIVNW LKKRTGPAAT TLPDGAAAES LVESSEVAVI GFFKDVESDS AKQFLQAAEA IDDIPFGITS NSDVFSKYQL DKDGVVLFKK FDEGRNNFEG EVTKENLLDF IKHNQLPLVI EFTEQTAPKI FGGEIKTHIL LFLPKSVSDY DGKLSNFKTA AESFKGKILF IFIDSDHTDN QRILEFFGLK KEECP | |
Greater than 90% as determined by SDS-PAGE. |
5034 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |