missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PARK7 / DJ-1 Protein
A DNA sequence encoding the Homo sapiens (Human) PARK7 / DJ-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8031-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001116849.1 | |
PARK7 / DJ-1 Protein | |
Research Use Only | |
PARK7 | |
Recombinant Protein | |
E. coli | |
MASKRALVIL AKGAEEMETV IPVDVMRRAG IKVTVAGLAG KDPVQCSRDV VICPDASLED AKKEGPYDVV VLPGGNLGAQ NLSESAAVKE ILKEQENRKG LIAAICAGPT ALLAHEIGFG SKVTTHPLAK DKMMNGGHYT YSENRVEKDG LILTSRGPGT SFEFALAIVE ALNG | |
Greater than 90% as determined by SDS-PAGE. |
11315 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |