missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCBP1 Protein
A cDNA sequence encoding the PCBP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12972-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PCBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDAGVTE SGLNVTLTIR LLMHGKEVGS IIGKKGESVK RIREESGARI NISEGNCPER IITLTGPTNA IFKAFAMIID KLEEDINSSM TNSTAASRPP VTLRLVVPAT QCGSLIGKGG CKIKEIREST GAQVQVAGDM LPNSTERAIT IAGVPQSVTE CVKQIC | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PCBP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |