missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDE6H Protein
A cDNA sequence encoding the PDE6H was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12991-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
5149 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
PDE6H | |
Human | |
His | |
PDE6H protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 2mM DTT and 0.1mM PMSF. |
PDE6H Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSDNTTL PAPASNQGPT TPRKGPPKFK QRQTRQFKSK PPKKGVKGFG DDIPGMEGLG TDITVICPWE AFSHLELHEL AQFGII |