missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDRG1 / C20orf126 Protein
A DNA sequence encoding the Homo sapiens (Human) PDRG1 / C20orf126, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8155-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_110442.1 | |
PDRG1 / C20orf126 Protein | |
Research Use Only | |
PDRG1 | |
Recombinant Protein | |
E. coli | |
MLSPEAERVL RYLVEVEELA EEVLADKRQI VDLDTKRNQN REGLRALQKD LSLSEDVMVC FGNMFIKMPH PETKEMIEKD QDHLDKEIEK LRKQLKVKVN RLFEAQGKPE LKGFNLNPLN QDELKALKVI LKG | |
Greater than 90% as determined by SDS-PAGE. |
81572 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |