missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFN2 Protein
A cDNA sequence encoding the PFN2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13021-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PFN2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAGWQSYVDN LMCDGCCQEA AIVGYCDAKY VWAATAGGVF QSITPIEIDM IVGKDREGFF TNGLALGAKK CSVIRDSLYV DGDCTMDIRT KSQGGEPTYN VAVGRAGRVL VFVMGKEGVH GGGLNKKAYS MAKYLRDSGF | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PFN2 protein solution (1 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |