missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphoacetylglucosamine mutase Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphoacetylglucosamine mutase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6487-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001186846.1 | |
Phosphoacetylglucosamine mutase Protein | |
Research Use Only | |
PGM3 | |
Recombinant Protein | |
E. coli | |
MDLGAITKYS ALHAKPNGLI LQYGTAGFRT KAEHLDHVMF RMGLLAVLRS KQTKSTIGVM VTASHNPEED NGVKLVDPLG EMLAPSWEEH ATCLANAEEQ DMQRVLIDIS EKEAVNLQQD AFVVIGRDTR PSSEKLSQSV IDGVTVLGGQ FHDYGLLTTP QLHYMVYCRN TGGRYGKATI EGYYQKLSKA FVELTKQASC SGDEYRSLKV DCANGIGALK LREMEHYFSQ GLSVQLFNDG SKGKLNHLCG ADFVKSHQKP PQGMEIKSNE RCCSFDGDAD RIVYYYHDAD GHFHLIDGDK IATLISSFLK ELLVEIGESL NIGVVQTAYA NGSSTRYLEE VMKVPVYCTK TGVKHLHHKA QEFDIGVYFE ANGHGTALFS TAVEMKIKQS AEQLEDKKRK AAKMLENIID LFNQAAGDAI SDMLVIEAIL ALKGLTVQQW DALYTDLPNR QLKVQVADRR VISTTDAERQ AVTPPGLQEA INDLVKKYKL SRAFVRPSGT EDVVRVYAEA DSQESADHLA HEVSLAVFQL AGGIGERPQP GF | |
Greater than 90% as determined by SDS-PAGE. |
5238 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |