missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIXY321 Protein
A cDNA sequence encoding the PIXY321 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10822-10ug
Additional Details : Peso : 0.01000kg
Specifications
PIXY321 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized after extensive dialysis against 10mM PBS, pH= 7.4.±0.1 and 0.1% BSA. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
Greater than 95.0% as determined by1. Analysis by RP-HPLC. 2. Analysis by SDS-PAGE. |