missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PKAR-I alpha Protein
A cDNA sequence encoding the PKAR-I alpha was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13062-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
5573 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MESGSTAASE EARSLRECEL YVQKHNIQAL LKDSIVQLCT ARPERPMAFL REYFERLEKE EAKQIQNLQK AGTRTDSRED EISPPPPNPV VKGRRRRGAI SAEVYTEEDA ASYVRKVIPK DYKTMAALAK AIEKNVLFSH LDDNERSDIF DAMFSVSFIA GETVIQQGDE GDNFYVIDQG ETDVYVNNEW ATSVGEGGSF GELALIYGTP RAATVKAKTN VKLWGIDRDS YRRILMGSTL RKRKMYEEFL SKVSILESLD KWERLTVADA LEPVQFEDGQ KIVVQGEPGD EFFIILEGSA AVLQRRSENE EFVEVGRLGP SDYFGEIALL MNRPRAATVV ARGPLKCVKL DRPRFERVLG PCSDILKRNI QQYNSFVSLS V | |
Greater than 95% as determined by SDS-PAGE. |
PKAR-I alpha Protein | |
Research Use Only | |
PRKAR1A | |
Human | |
Untagged | |
PKA regulatory subunit-I alpha is supplied at a concentration of 1.3 mg/ml in 20mM MOPS (pH 7.0), 150mM NaCl, 1mM 2-mercaptoethanol and 50% glycerol. |