missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plasminogen Protein
A DNA sequence encoding the Human Plasminogen, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10364-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000292.1 | |
Plasminogen Protein | |
Research Use Only | |
PLG | |
The specific activity determined by an assay on anti-proliferation and anti-migration using endothelial cells in vitro and anti-angiogenesis in vivo is 5.5 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM NaAc, pH 5.5, 4 % mannitol, lyophilized |
5340 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VYLSECKTGN GKNYRGTMSK TKNGITCQKW SSTSPHRPRF SPATHPSEGL EENYCRNPDN DPQGPWCYTT DPEKRYDYCD ILECEEECMH CSGENYDGKI SKTMSGLECQ AWDSQSPHAH GYIPSKFPNK NLKKNYCRNP DRELRPWCFT TDPNKRWELC DIPRCTTPPP SSGPTYQCLK GTGENYRGNV AVTVSGHTCQ HWSAQTPHTH NRTPENFPCK NLDENYCRNP DGKRAPWCHT TNSQVRWEYC KIPSCDSSP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |