missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLAUR / CD87 / uPAR Protein
A DNA sequence encoding the Homo sapiens (Human) PLAUR / CD87 / uPAR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6503-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001005376.1 | |
PLAUR / CD87 / uPAR Protein | |
Research Use Only | |
PLAUR | |
Recombinant Protein | |
E. coli | |
LRCMQCKTNG DCRVEECALG QDLCRTTIVR LWEEGEELEL VEKSCTHSEK TNRTLSYRTG LKITSLTEVV CGLDLCNQGN SGRAVTYSRS RYLECISCGS SDMSCERGRH QSLQCRSPEE QCLDVVTHWI QEGEEGRPKD DRHLRGCGYL PGCPGSNGFH NNDTFHFLKC CNTTKCNEGP ILELENLPQN GRQCYSCKGN STHGCSSEET FLIDCRGPMN QCLVATGTHE PKNQSYMVRG CATASMCQHA HLGDAFSMNH IDVSCCTKSG CNHPDLDVQY RSG | |
Greater than 90% as determined by SDS-PAGE. |
5329 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |