missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plexin-A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Plexin-A1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8790-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_115618.3 | |
Plexin-A1 Protein | |
Research Use Only | |
PLXNA1 | |
Recombinant Protein | |
E. coli | |
RAGGGSQPPF RTFSASDWGL THLVVHEQTG EVYVGAVNRI YKLSGNLTLL RAHVTGPVED NEKCYPPPSV QSCPHGLGST DNVNKLLLLD YAANRLLACG SASQGICQFL RLDDLFKLGE PHHRKEHYLS SVQEAGSMAG VLIAGPPGQG QAKLFVGTPI DGKSEYFPTL SSRRLMANEE DADMFGFVYQ DEFVSSQLKI PSDTLSKFPA FDIYYVYSFR SEQFVYYLTL QLDTQLTSPD AAGEHFFTSK IVRLCVDDPK FYSYV | |
Greater than 90% as determined by SDS-PAGE. |
5361 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |