missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plexin-A4 Protein
A DNA sequence encoding the Homo sapiens (Human) Plexin-A4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8056-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001099013.1 | |
Plexin-A4 Protein | |
Research Use Only | |
PLXNA4 | |
Recombinant Protein | |
E. coli | |
TLLTRQPAPL SQKQRSFVTF RGEPAEGFNH LVVDERTGHI YLGAVNRIYK LSSDLKVLVT HETGPDEDNP KCYPPRIVQT CNEPLTTTNN VNKMLLIDYK ENRLIACGSL YQGICKLLRL EDLFKLGEPY HKKEHYLSGV NESGSVFGVI VSYSNLDDKL FIATAVDGKP EYFPTISSRK LTKNSEADGM FAYVFHDEFV ASMIKIPSDT FTIIPDFDIY YVYGFSSGNF VYFLTLQPEM VSPPGSTTKE QVYTSKLVRL CKEDTAFNSY VEVPIGCERS GVEYRLLQAA YLSKAGAVLG RTLGVHPDDD LLFTVFSKGQ KRKMKSLDES ALCIFILKQI NDRIKERLQS CYRGEGTLDL AWLKVKDIPC SSALLTIDDN FCGLDMNAPL GVSDMVRGIP VFTEDRDRMT SVIAYVYKNH SLAFVGTKSG KLKKIRVDGP RGNALQYETV QVVDPGPVLR DMAFSKDHEQ LYIMSERQLT RVPVESCGQY QSCGECLGS | |
Greater than 90% as determined by SDS-PAGE. |
91584 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |