missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PON2 Protein
A cDNA sequence encoding the PON2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13110-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PON2 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PON2 is supplied in PBS and 50% glycerol. |
Greater than 95% as determined by SDS-PAGE. Single band on Western Blot. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGRLVAVGLLGIALALLGERLLALRNRLKASREVESVDLPHCHLIKGIEAGSEDID ILPNGLAFFSVGLKFPGLHSFAPDKPGGILMMDLKEEKPRARELRISRGFDLASFNP HGISTFIDNDDTVYLFVVNHPEFKNTVEIFKFEEAENSLLHLKTVKHELLPSVNDIT AVGPAHFYATNDHYFSDPFLKYLETYLNLHWANVVYYSPNEVKVVAEGFDSAN GINISPDDKYIYVADILAHEIHVLEKHTNMNLTQLKVLELDTLVDNLSIDPSSGDIW VGCHPNGQKLFVYDPNNPPSSEVLRIQNILSEKPTVTTVYANNGSVLQGSSVASVY DGKLLIGTLYHRALYCELZ |