missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POU domain, class 5, transcription factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) POU domain, class 5, transcription factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6517-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001272916.1 | |
POU domain, class 5, transcription factor 1 Protein | |
Research Use Only | |
POU5F1 | |
Recombinant Protein | |
E. coli | |
MAGHLASDFA FSPPPGGGGD GPGGPEPGWV DPRTWLSFQG PPGGPGIGPG VGPGSEVWGI PPCPPPYEFC GGMAYCGPQV GVGLVPQGGL ETSQPEGEAG VGVESNSDGA SPEPCTVTPG AVKLEKEKLE QNPEESQDIK ALQKELEQFA KLLKQKRITL GYTQADVGLT LGVLFGKVFS QTTICRFEAL QLSFKNMCKL RPLLQKWVEE ADNNENLQEI CKAETLVQAR KRKRTSIENR VRGNLENLFL QCPKPTLQQI SHIAQQLGLE KDVVRVWFCN RRQKGKRSSS DYAQREDFEA AGSPFSGGPV SFPLAPGPHF GTPGYGSPHF TALYSSVPFP EGEAFPPVSV TTLGSPMHSN | |
Greater than 80% as determined by SDS-PAGE. |
5460 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |