missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRAP1 Protein
A cDNA sequence encoding the PRAP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13133-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PRAP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVPAPKVPIKM QVKHWPSEQD PEKAWGARVV EPPEKDDQLV VLFPVQKPKL LTTEEKPRGQ GRGPILPGTK AWMETEDTLG HVLSPEPDHD SLYHPPPEED QGEERPRLWV MPNHQVLLGP EEDQDHIYHP Q | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRAP1 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |