missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRH1 Protein
A cDNA sequence encoding the PRH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13137-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PRH1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQDLNEDV SQEDVPLVIS DGGDSEQFLD EERQGPPLGG QQSQPSAGDG NQDDGPQQGP PQQGGQQQQG PPPPQGKPQG PPQQGGHPPP PQGRPQGPPQ QGGHPRPPR | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PRH1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |