missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRND Protein
A cDNA sequence encoding the PRND was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13149-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
23627 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
PRND | |
Human | |
His | |
PRND protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 30% glycerol and 1mM DTT. |
PRND Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRGIKHRI KWNRKALPST AQITEAQVAE NRPGAFIKQG RKLDIDFGAE GNRYYEANYW QFPDGIHYNG CSEANVTKEA FVTGCINATQ AANQGEFQKP DNKLHQQVLW RLVQELCSLK HCEFWLERG |