missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prolactin-inducible Protein
A DNA sequence encoding the Homo sapiens (Human) Prolactin-inducible protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6495-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002643.1 | |
Prolactin-inducible Protein | |
Research Use Only | |
Pip | |
Recombinant Protein | |
E. coli | |
QDNTRKIIIK NFDIPKSVRP NDEVTAVLAV QTELKECMVV KTYLISSIPL QGAFNYKYTA CLCDDNPKTF YWDFYTNRTV QIAAVVDVIR ELGICPDDAA VIPIKNNRFY TIEILKVE | |
Greater than 90% as determined by SDS-PAGE. |
5304 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |