missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PROS1 / Protein
A DNA sequence encoding the Homo sapiens (Human) PROS1 /, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6533-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000304.2 | |
PROS1 / Protein | |
Research Use Only | |
PROS1 | |
Recombinant Protein | |
E. coli | |
ANSLLEETKQ GNLERECIEE LCNKEEAREV FENDPETDYF YPKYLVCLRS FQTGLFTAAR QSTNAYPDLR SCVNAIPDQC SPLPCNEDGY MSCKDGKASF TCTCKPGWQG EKCEFDINEC KDPSNINGGC SQICDNTPGS YHCSCKNGFV MLSNKKDCKD VDECSLKPSI CGTAVCKNIP GDFECECPEG YRYNLKSKSC EDIDECSENM CAQLCVNYPG GYTCYCDGKK GFKLAQDQKS CEVVSVCLPL NLDTKYELLY LAEQFAGVVL YLKFRLPEIS RFSAEFDFRT YDSEGVILYA ESIDHSAWLL IALRGGKIEV QLKNEHTSKI TTGGDVINNG LWNMVSVEEL EHSISIKIAK EAVMDINKPG PLFKPENGLL ETKVYFAGFP RKVESELIKP INPRLDGCIR SWNLMKQGAS GIKEIIQEKQ NKHCLVTVEK GSYYPGSGIA QFHIDYNNVS SAEGWHVNVT LNIRPSTGTG VMLALVSGNN TVPFAVSLVD STSEKSQDIL LSVENTVIYR IQALSLCSDQ QSHLEFRVNR NNLELSTPLK IETISHEDLQ RQLAVLDKAM KAKVATYLGG LPDVPFSATP VNAFYNGCME VNINGVQLDL DEAISKHNDI RAHSCPSVWK KTKNS | |
Greater than 90% as determined by SDS-PAGE. |
5627 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |