missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prostate stem cell antigen Protein
A DNA sequence encoding the Homo sapiens (Human) Prostate stem cell antigen, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8733-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Prostate stem cell antigen Protein | |
Research Use Only | |
PSCA | |
Recombinant Protein | |
Yeast | |
LLCYSCKAQV SNEDCLQVEN CTQLGEQCWT ARIRAVGLLT VISKGCSLNC VDDSQDYYVG KKNITCCDTD LCNAS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |