missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit alpha type-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit alpha type-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6543-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002777.1 | |
Proteasome subunit alpha type-1 Protein | |
Research Use Only | |
PSMA1 | |
Recombinant Protein | |
E. coli | |
MFRNQYDNDV TVWSPQGRIH QIEYAMEAVK QGSATVGLKS KTHAVLVALK RAQSELAAHQ KKILHVDNHI GISIAGLTAD ARLLCNFMRQ ECLDSRFVFD RPLPVSRLVS LIGSKTQIPT QRYGRRPYGV GLLIAGYDDM GPHIFQTCPS ANYFDCRAMS IGARSQSART YLERHMSEFM ECNLNELVKH GLRALRETLP AEQDLTTKNV SIGIVGKDLE FTIYDDDDVS PFLEGLEERP QRKAQPAQPA D | |
Greater than 90% as determined by SDS-PAGE. |
5682 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |