missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein C8orf4 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein C8orf4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8178-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_064515.1 | |
Protein C8orf4 Protein | |
Research Use Only | |
C8orf4 | |
Recombinant Protein | |
E. coli | |
MKAKRSHQAV IMSTSLRVSP SIHGYHFDTA SRKKAVGNIF ENTDQESLER LFRNSGDKKA EERAKIIFAI DQDVEEKTRA LMALKKRTKD KLFQFLKLRK YSIKVH | |
Greater than 80% as determined by SDS-PAGE. |
56892 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |