missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein prune homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Protein prune homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7790-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001290158.1 | |
Protein prune homolog Protein | |
Research Use Only | |
PRUNE | |
Recombinant Protein | |
E. coli | |
MLRKDQKTIY RQGVKVAISA IYMDLEICEV LERSHSPPLK LTPASSTHPN LHAYLQGNTQ VSRKKLLPLL QEALSAYFDS MKIPSGQPET ADVSREQVDK ELDRASNSLI SGLSQDEEDP PLPPTPMNSL VDECPLDQGL PKLSAEAVFE KCSQISLSQS TTASLSKK | |
Greater than 90% as determined by SDS-PAGE. |
58497 | |
10 μg | |
Not determined. | |
Full Length of Isoform 6 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |