missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTGDS / L-PGDS Protein
A DNA sequence encoding the Homo sapiens (Human) PTGDS / L-PGDS, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6550-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000945.3 | |
PTGDS / L-PGDS Protein | |
Research Use Only | |
PTGDS | |
Recombinant Protein | |
E. coli | |
APEAQVSVQP NFQQDKFLGR WFSAGLASNS SWLREKKAAL SMCKSVVAPA TDGGLNLTST FLRKNQCETR TMLLQPAGSL GSYSYRSPHW GSTYSVSVVE TDYDQYALLY SQGSKGPGED FRMATLYSRT QTPRAELKEK FTAFCKAQGF TEDTIVFLPQ TDKCMTEQ | |
Greater than 86% as determined by SDS-PAGE. |
5730 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |