missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTP4A1 Protein
A cDNA sequence encoding the PTP4A1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13208-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PTP4A1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVTYKNMRFL ITHNPTNATL NKFIEELKKY GVTTIVRVCE ATYDTTLVEK EGIHVLDWPF DDGAPPSNQI VDDWLSLVKI KFREEPGCCI AVHCVAGLGR APVLVALALI EGGMKYEDAV QFIRQKRRGA FNSKQLLYLE KYRPKMRLRF KDSNGHRNNC | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PTP4A1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |