missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRN Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9748-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001186692.1 | |
PTPRN Protein | |
Research Use Only | |
PTPRN | |
Recombinant Protein | |
Yeast | |
VSAHGCLFDR RLCSHLEVCI QDGLFGQCQV GVGQARPLLQ VTSPVLQRLQ GVLRQLMSQG LSWHDDLTQY VISQEMERIP RLRPPEPRPR DRSGLAPKRP GPAGELLLQD IPTGSAPAAQ HRLPQPPVGK GGAGASSSLS PLQAELLPPL LEHLLLPPQP PHPSLSYEPA LLQPYLFHQF GSRDGSRVSE GSPGMVSVGP LPKAEAPALF SRTASKGIFG DHPGHSYGDL PGPSPAQLFQ DSGLLYLAQE LPAPSRARVP RLPEQGSSSR AEDSPEGYEK EGLGDRGEKP ASPAVQPDAA LQRLAAVLAG YGVELRQLTP EQLSTLLTLL QLLPKGAGRN PGGVVNVGAD IKKTMEGPVE GRDTAELPAR TSPMPGHPTA SPTSSEVQQV PSPVSSEPPK AARPPVTPVL LEKKSPLGQS QPTVAGQPSA RPAAEEYGYI VTDQKPLSLA AGVKLLEILA EHVHMSSGSF INISVVGPAL TFRIRHNEQN LSLADVTQQA GLVKSELEAQ TGLQILQTGV GQREEAAAVL PQTAHSTSPM R | |
Greater than 90% as determined by SDS-PAGE. |
5798 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |