missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRZ1 Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRZ1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6557-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001193767.1 | |
PTPRZ1 Protein | |
Research Use Only | |
PTPRZ1 | |
Recombinant Protein | |
Yeast | |
IGWSYTGALN QKNWGKKYPT CNSPKQSPIN IDEDLTQVNV NLKKLKFQGW DKTSLENTFI HNTGKTVEIN LTNDYRVSGG VSEMVFKASK ITFHWGKCNM SSDGSEHSLE GQKFPLEMQI YCFDADRFSS FEEAVKGKGK LRALSILFEV GTEENLDFKA IIDGVESVSR FGKQAALDPF ILLNLLPNST DKYYIYNGSL TSPPCTDTVD WIVFKDTVSI SESQLAVFCE VLTMQQSGYV MLMDYLQNNF REQQYKFSRQ VFSSY | |
Greater than 90% as determined by SDS-PAGE. |
5803 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |