missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PURB Protein
A cDNA sequence encoding the PURB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13218-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PURB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADGDSG SERGGGGGPC GFQPASRGGG EQETQELASK RLDIQNKRFY LDVKQNAKGR FLKIAEVGAG GSKSRLTLSM AVAAEFRDSL GDFIEHYAQL GPSSPEQLAA GAEEGGGPRR ALKSEFLVRE NRKYYLDLKE NQRGRFLRIR QTVNRGGGGF GAGPGPGGLQ SGQTIALPAQ GLIEFRDALA KLIDDYGGED DELAGGPGGG AGGPGGGLYG ELPEGTSITV DSKRFFFDVG CNKYGVFLRV SEVKPSYRNA ITVPFKAWGK FGGAFCRYAD EMKEIQERQR DKLYERRGGG SGGGEESEGE EVDED | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PURB solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |