missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Putative inactive neutral ceramidase B Protein
A DNA sequence encoding the Homo sapiens (Human) Putative inactive neutral ceramidase B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5682-ye-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001072984.1 | |
Putative inactive neutral ceramidase B Protein | |
Research Use Only | |
ASAH2B | |
Recombinant Protein | |
Yeast | |
MRQHRQFMDR THYLLTFSSS ETLLRLLLRI VDRAPKGRTF GDVLQPAKPE YRVGEVAEVI FVGANPKNSV QNQTHQTFLT VEKYEATSTS WQIVCNDASW ETRFYWHKGL LGLSNATVEW HIPDTAQPGI YRIRYFGHNR KQDILKPAVI LSFEGTSPAF EVVTI | |
Greater than 90% as determined by SDS-PAGE. |
653308 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |